Alfa Aesar™ Trypsin Inhibitor, chicken egg whites

Inhibits the pancreatic enzyme trypsin

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J63927.03

1GR Trypsin inhibitor, chicken egg whites

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: CAS Number: 9035-81-8 Weight: 0.00100kg



Product Code. 15426089

Quantity Price
1 £ 105.15 / 1g
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Specification

CAS 9035-81-8
Synonym Trypsin inhibitor, ovomucoid
Quantity 1g
Product Type Signal Transduction Reagent
Description White
Format Powder
For Use With (Application) Inhibits the pancreatic enzyme trypsin