missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Trypsin (Bovine), Sequencing Grade

Used for Enzyme

Brand:  Abnova™ P5319.4x25ug

Additional Details : Weight : 0.00010kg

Product Code. 16141840

  • £125.00 / Pack of 4
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

#REF!
Specifications

Specifications

Q29463
282603/615237
Native protein purified from Bovine Pancreas
Store at -20°C on dry atmosphere.
TRYP8
PRSS2
Bovine
Native protein purified from Bovine Pancreas
Enzyme Activity
Trypsin (Bovine), Sequencing Grade
4 x 25 ug
RUO
PRSS2
≥150 TAME units per mg protein (at least 8,625 BAEE/2875 USP/NF units per mg protein). TAME Unit: One Unit hydrolyzes 1 umole of p-toluene-sulfonyl-L-arginine methyl ester (TAME) per minute at 25°C, pH 8.2, in the presence of 10mM calcium.
None
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers