Learn More
Alfa Aesar™ Trypsin, bovine pancreas
A pancreatic serine protease
Brand: Alfa Aesar™ J63688.09
Code : NEW
Additional Details : CAS Number : 9002-07-7 Weight : 0.01000kg
Specifications
Powder | |
A pancreatic serine protease | |
10g | |
9002-07-7 |
White to Off-white | |
Trypsin | |
−30° to −10°C | |
EC 3.4.21.4; Peptidyl peptide hydrolase |