missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Trypsin 1/PRSS1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Trypsin 1/PRSS1 |
---|---|
Clone | 446628 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Trypsin 1/PRSS1 Monoclonal specifically detects Trypsin 1/PRSS1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Trypsin 1/PRSS1 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P07477 | |
5644 | |
Mouse myeloma cell line NS0-derived recombinant human Trypsin 1/PRSS1 Ala16-Ser247 Accession # P07477 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Trpysin 1/PRSS1 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse (rm) Trypsin 3, recombinant human (rh) Trypsin 2, or rhTrypsin 3. |
446628 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Beta-trypsin, Cationic trypsinogen, digestive zymogen, DKFZp779A0837, EC 3.4.21, EC 3.4.21.4, FLJ57296, MGC120175, MGC149362, nonfunctional trypsin 1, protease, serine, 1 (trypsin 1), PRSS1, PTRYI, Serine protease 1, TRP1, TRY1beta-trypsin, TRY4, Trygn16, TRYP1, Trypsin I, trypsin-1, trypsinogen 1, trypsinogen A | |
PRSS1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only