missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trypsin 1:250, porcine pancreas

A pancreatic serine protease

Brand:  Alfa Aesar™ J63993.09

UNSPSC : 12352204

Code : NEW

Additional Details : CAS Number : 9002-07-7 Weight : 0.01000kg

 View more versions of this product

Product Code. 15426139

  • £80.56 / 10g


Estimated Shipment date
from Supplier 06-09-2017
Add to basket

Specifications

Specifications

Powder
A pancreatic serine protease
10g
9002-07-7
White to Off-White
Trypsin
−30° to −10°C
EC 3.4.21.4
Documentation

Technical Documents