missing translation for 'onlineSavingsMsg'
Learn More

TrueStart Hot Start Taq DNA Polymerase

Improve specificity and sensitivity of hot-start PCR with this polymerase.

Brand:  Thermo Scientific EP0613

Code : LD

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11561515

  • £311.00 / Pack of 5


Estimated Shipment
01-02-2018
Add to basket

Disclaimers:

In certain countries use of this product is covered by patents. Purchase of product in these countries includes non-transferable, limited license for using only this amount of product for the purchaser's own internal research.

This product or the use of this product is covered by US patent application US20070224611A1 and corresponding counterparts owned by Fermentas. The purchase of this product includes a non-transferable license to use this product for the purchaser's internal research. All other commercial uses of this product, including without limitation product use for diagnostic purposes, resale of product in the original or any modified form or product use in providing commercial services require a separate license from Fermentas.


Description

Description

Highlights

  • Fastest activating hot start Taq DNA polymerase
  • High PCR specificity -reduced effects of mis-priming and primer-dimer formation
  • Enhanced PCR sensitivity
  • Convenient room temperature PCR set-up
  • Generates PCR products with 3'-dA overhangs
  • Specifications

    Specifications

    High throughput hot start PCR; RT-PCR; Highly specific amplification of genomic and cDNA targets up to 3kb; Amplification of low copy DNA targets; Real-time PCR; Mul
    10 x 1.25mL 10X TrueStart Taq Buffer and 10 x 1.25mL 25mM MgCl2
    TrueStart Hot Start Taq DNA Polymerase
    5 x 500U
    Videos