missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPC7 Monoclonal Antibody (N64A/36), PerCP, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA545475
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45475 was sufficient for detection of TrpC7 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.∣ Detects approximately 100kDa.
TRPC7 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC7 is probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. TRPC7 is also activated by diacylglycerol (DAG). Further, TRPC7 may also be activated by intracellular calcium store depletion.Specifications
TRPC7 | |
Monoclonal | |
1 mg/mL | |
2.15mM MES, 47.35mM phosphate with 1.033mM EDTA and no preservative; pH 7.4 | |
Q9HCX4, Q9WVC5 | |
TRPC7 | |
Synthetic peptide amino acids 845-862 of human TRPC7 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Microarray, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
N64A/36 | |
PerCP | |
TRPC7 | |
capacitative calcium channel TRPC7; hTRP7; likley ortholog of mouse transient receptor potential cation channel, subfamily C, member 7; mTRP7; putative capacitative calcium channel; short transient receptor potential channel 7; transient receptor potential cation channel subfamily C member 7; transient receptor potential cation channel, subfamily C, member 7; transient receptor protein 7; TRP7; TRP-7; TRP7 protein; TRPC7; Trrp8 | |
Mouse | |
Protein G | |
RUO | |
26946, 282822, 57113 | |
4° C | |
IgG1 |