missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPC4 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111757
This item is not returnable.
View return policy
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)KYSALNP RESWD Homology: Mouse, human amino acid residues are identical.
TRPC4 is a member of the canonical subfamily of transient receptor potential cation channels. TRPC4 forms a non-selective calcium-permeable cation channel that is activated by Gq-coupled receptors and tyrosine kinases, and plays a role in multiple processes including endothelial permeability, vasodilation, neurotransmitter release and cell proliferation. Single nucleotide polymorphisms in TRPC4 may be associated with generalized epilepsy with photosensitivity. Alternatively spliced transcript variants encoding multiple isoforms have been observed for TRPC4. The classical or canonical transient receptor potential (TRPC) channels are the subfamily most closely related to the founding member of the TRP family, the Drosophila TRP channel. TRPC channels have seven TRPC genes that encode the channel protein (TRPC1-7). Diseases associated with TRPC4 include Photosensitive Epilepsy.CaV3.3 encodes the pore-forming alpha subunit of a voltage gated calcium channel. The encoded protein is a member of a subfamily of calcium channels referred to as is a low voltage-activated, T-type, calcium channel. The channel encoded by CaV3.3 is characterized by a slower activation and inactivation compared to other T-type calcium channels. CaV3.3 may be involved in calcium signaling in neurons. Alternate splicing results in multiple transcript variants of CaV3.3. Voltage-gated calcium channels (CaV) are present in the membrane of most excitable cells and mediate calcium influx in response to depolarization, an proteins such as CaV3.3 regulate intracellular processes such as contraction, secretion, neurotransmission and gene expression. Diseases associated with CACNA1I include Childhood Absence Epilepsy.Specifications
TRPC4 (extracellular) | |
Polyclonal | |
Unconjugated | |
Trpc4 | |
alpha TRPC4; beta TRPC4; beta-2 TRPC4; Capacitative calcium entry channel Trp4; CCE1; HTRP4; hTrp-4; MGC119570; MGC119571; MGC119572; MGC119573; Receptor-activated cation channel TRP4; Short transient receptor potential channel 4; STRPC4; TRP4; TRPC 4; TRPC4; Trpc4_v2; Trp-related protein 4; Trrp4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22066, 7223, 84494 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunomicroscopy, Western Blot | |
0.85 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
O35119, Q9QUQ5, Q9UBN4 | |
Trpc4 | |
Peptide corresponding to amino acid residues 458-469 of rat TRPC4 (2ndextracellular loop) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |