missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tropine DL-tropate, 99%

Reduces brain levels of acetylcholine

Brand:  Alfa Aesar™ J64677.06

Code : NEW

Additional Details : CAS Number : 51-55-8 Weight : 0.00500kg Transport : UN number : 1544 Chem class : 2 Pack group : II

 View more versions of this product

Product Code. 15456519

  • £39.00 / 5g


Estimated Shipment date
from Supplier 23-01-2018
Add to basket

Specifications

Specifications

5g
Powder
0.99
C17H23NO3
DL-Hyoscyamine; Atropine
Signal Transduction Reagent
White
Reduces brain levels of acetylcholine
51-55-8
289.37g/mol
Light Sensitive
Videos
SDS