missing translation for 'onlineSavingsMsg'
Learn More

Tropine, 98%, Thermo Scientific Chemicals

Catalog Number p-3737709
34.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
120-29-6
C8H15NO
141.21
MFCD00005551
CYHOMWAPJJPNMW-RNFRBKRXSA-N
tropine, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, spectrum3_001840, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
6101956
(1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
CN1C2CCC1CC(C2)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
120-29-6
C8H15NO
141.21
MFCD00005551
CYHOMWAPJJPNMW-RNFRBKRXSA-N
tropine, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, spectrum3_001840, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
6101956
(1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
CN1C2CCC1CC(C2)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 62.0°C to 64.0°C
Color White to Yellow
Density 1.0400g/mL
Boiling Point 233.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 16
Merck Index 15, 9960
Specific Gravity 1.04
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in ether and chloroform, freely soluble in alcohol
Formula Weight 141.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tropine
Show More Show Less
compliance-icons
Product Identifier
  • Tropine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'