missing translation for 'onlineSavingsMsg'
Learn More

Troglitazone, Tocris Bioscience™

Selective PPARγ agonist; antidiabetic agent

Brand:  Tocris (Bio-Techne) 311410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13452464

  • £149.94 / 10mg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective PPARγ receptor agonist (EC50 values are 780 and 555 nM at murine and human PPARγ receptors respectively). Displays no activity at PPARα or PPARδ receptors. Antidiabetic agent; exerts potent glucose-lowering effects in insulin-resistant diabetic mice. Displays anti-invasive effect on human breast cancer cells; reduces migration, adhesion and spreading on fibronectin-coated plates. Also inhibits cell growth of hematopoietic cell lines. Inhibits lamellipodia formation and actin polymerization.
Specifications

Chemical Identifiers

97322-87-7
441.542
troglitazone, rezulin, romglizone, prelay, romozin, noscal, rezulin tn, troglitazona, troglitazonum, ccris 8969
CHEBI:9753
CC1=C(C(=C2CCC(OC2=C1C)(C)COC3=CC=C(C=C3)CC4C(=O)NC(=O)S4)C)O
C24H27NO5S
GXPHKUHSUJUWKP-UHFFFAOYSA-N
5591
5-[[4-[(6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydrochromen-2-yl)methoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione

Specifications

10mg
97322-87-7
troglitazone, rezulin, romglizone, prelay, romozin, noscal, rezulin tn, troglitazona, troglitazonum, ccris 8969
CC1=C(C(=C2CCC(OC2=C1C)(C)COC3=CC=C(C=C3)CC4C(=O)NC(=O)S4)C)O
441.542
CHEBI:9753
>97%
Troglitazone
C24H27NO5S
GXPHKUHSUJUWKP-UHFFFAOYSA-N
5-[[4-[(6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydrochromen-2-yl)methoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
5591
441.54
Videos
Documents