missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triton™ X-100, Molecular Biology Reagent Grade, MP Biomedicals™
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Specifications
Color | Colorless to Yellow |
Physical Form | Viscous Liquid |
Density | 8.9 lb./gal. |
Quantity | 50 mL |
Grade | Molecular Biology |
Chemical Name or Material | Triton™ X-100 |
Safety and Handling
Product Identifier
- Triton X-100, molecular biology reagent
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- H300-Fatal if swallowed.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : triton-X 100
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'