missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Triton|r X-100, Electrophoresis Reagent
Click to view available options
Quantity:
100 mL
2500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Content And Storage | Ambient temperatures |
Density | 1.061 g/mL |
Formula Weight | ca. 625 |
MDL Number | MFCD00132505 |
Quantity | 2500 mL |
Melting Point | ∼5°C |
Odor | Mild |
InChI Key | JYCQQPHGFMYQCF-UHFFFAOYSA-N |
SMILES | CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'