missing translation for 'onlineSavingsMsg'
Learn More

Tris(triphenylphosphine)rhodium(I) chloride, 98%, ACROS Organics™

£63.07 - £223.28

Chemical Identifiers

CAS 14694-95-2
Molecular Formula C54H45ClP3Rh
Molecular Weight (g/mol) 925.23
MDL Number MFCD00010016
InChI Key QBERHIJABFXGRZ-UHFFFAOYSA-M
Synonym chlorotris triphenylphosphine rhodium i, chlorotris triphenylphosphine rhodium, tris triphenylphosphine rhodium i chloride, tris triphenylphosphine chlororhodium, wilkinson's catalyst, unii-0fv534bkev, tris triphenylphosphine rhodium chloride, 0fv534bkev, rhodium; triphenylphosphane; chloride
PubChem CID 84599
IUPAC Name rhodium;triphenylphosphane;chloride
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Rh]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10793191
SDS View Documents
Acros Organics
190420010
1g Glass bottle £63.07
1g View alternative products
Save Up to 
»
In stock, ready to ship: 13 Please to purchase this item. Need a web account? Register with us today!
10104260
SDS View Documents
Acros Organics
190420050
5g Glass bottle £223.28
5g View alternative products
Save Up to 
»
In stock, ready to ship: 4 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reduction reagent
Specifications

Chemical Identifiers

14694-95-2
925.23
QBERHIJABFXGRZ-UHFFFAOYSA-M
84599
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Rh]
C54H45ClP3Rh
MFCD00010016
chlorotris triphenylphosphine rhodium i, chlorotris triphenylphosphine rhodium, tris triphenylphosphine rhodium i chloride, tris triphenylphosphine chlororhodium, wilkinson's catalyst, unii-0fv534bkev, tris triphenylphosphine rhodium chloride, 0fv534bkev, rhodium; triphenylphosphane; chloride
rhodium;triphenylphosphane;chloride

Specifications

Tris(triphenylphosphine)rhodium(I) chloride
Glass bottle
245.0°C to 250.0°C
98%
98%
[(C6H5)3P]3RhCl
01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,90
chlorotris triphenylphosphine rhodium i, chlorotris triphenylphosphine rhodium, tris triphenylphosphine rhodium i chloride, tris triphenylphosphine chlororhodium, wilkinson's catalyst, unii-0fv534bkev, tris triphenylphosphine rhodium chloride, 0fv534bkev, rhodium; triphenylphosphane; chloride
QBERHIJABFXGRZ-UHFFFAOYSA-M
rhodium;triphenylphosphane;chloride
84599
Crystalline Powder
Authentic
Brown-Red to Maroon
1g
14694-95-2
C54H45ClP3Rh
MFCD00010016
15, 10245
Solubility in water: insoluble. Other solubilities: soluble in chloroform, dichloromethane, benzene,, toluene, less soluble in acetic acid, ketones, methanol and, lower aliphatic alcohols, virtually insoluble in alkanes and cyclohexane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Rh]
925.23
925.23
98%