missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyloxy)ethylene, 95%, ACROS Organics™

Brand:  Acros Organics 216010050

Code : 29

Additional Details : CAS Number : 69097-20-7 Weight : 0.05500kg Transport : UN number : 1993 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10707022

  • £52.30 / 5g


Estimated Shipment
22-05-2018
Add to basket


Specifications

Chemical Identifiers

Tris(trimethylsilyloxy)ethylene
95%
[(CH3)3SiO]2C=CHOSi(CH3)3
MFCD00011641
95%
C11H28O3Si3
292.59

Specifications

Tris(trimethylsilyloxy)ethylene
Authentic
95%
C11H28O3Si3
Glass bottle
MFCD00011641
1.4190 to 1.4220
0.885
Clear colorless to yellow
39°C
69097-20-7
Liquid
95%
[(CH3)3SiO]2C=CHOSi(CH3)3
292.59
08,523; 09,512
Solubility in water: insoluble.
0.8850g/mL
90.0°C (1.0 mmHg)
5g
Videos
SDS