missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl)silane, 96%, Thermo Scientific Chemicals

Catalog Number p-3779842
Click to view available options
Quantity:
2.5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1873-77-4
C9H28Si4
248.66
MFCD00077893
SQMFULTZZQBFBM-UHFFFAOYSA-N
tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl
1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C
This item is not returnable. View return policy
1873-77-4
C9H28Si4
248.66
MFCD00077893
SQMFULTZZQBFBM-UHFFFAOYSA-N
tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl
1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Boiling Point 82°C to 84°C (12.0mmHg)
Color Colorless to Yellow
Physical Form Liquid
Quantity 2.5 mL
Assay Percent Range 95% min. (GC)
Linear Formula ((CH3)3Si)3SiH
Fieser 15,358; 17,395
Flash Point 55°C
Merck Index 15, 9934
Solubility Information Solubility in water: insoluble
Formula Weight 248.66
Percent Purity 96%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4890 to 1.491
Specific Gravity 0.8
Density 0.8000g/mL
Chemical Name or Material Tris(trimethylsilyl)silane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tris(trimethylsilyl)silane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'