missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl)phosphine, Thermo Scientific™

Catalog Number 11480510
Click to view available options
1g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 15573-38-3
Density 0.863 g/mL
Boiling Point 102°C to 105°C (16mmHg)
Odor Pungent
Molecular Formula C9H27PSi3
Refractive Index 1.502
MDL Number MFCD00015487
Quantity 1g
UN Number UN2845
Sensitivity Air and moisture sensitive
Synonym tris trimethylsilyl phosphine, tris trimethylsilyl phosphane, phosphine, tris trimethylsilyl, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl disilaphosphane, tris trimethylsilyl phosphine in hexane, pubchem6456, acmc-1btki, tris-trimethylsilyl phosphine, phosphine,tris trimethylsilyl
Solubility Information Insoluble in water.
InChI Key OUMZKMRZMVDEOF-UHFFFAOYSA-N
SMILES C[Si](C)(C)P([Si](C)(C)C)[Si](C)(C)C
IUPAC Name tris(trimethylsilyl)phosphane
Molecular Weight (g/mol) 250.544
PubChem CID 272683
Formula Weight 250.55
Chemical Name or Material Tris(trimethylsilyl)phosphine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'