missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(trimethylsilyl)amine, 99%, Thermo Scientific Chemicals
23.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 67°C to 69°C |
Color | Colorless |
Boiling Point | 72°C (10.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | N(Si(CH3)3)3 |
Quantity | 5 g |
Synonym | tris trimethylsilyl amine, amine, tris trimethylsilyl, silanamine, 1,1,1-trimethyl-n,n-bis trimethylsilyl, silanamine,1,1,1-trimethyl-n,n-bis trimethylsilyl, nitrilotris trimethylsilane, ch3 3si 3n, n,n,n-tris trimethylsilyl amine #, bis trimethylsilyl amino-dimethylsilyl methane |
Solubility Information | Solubility in water: insoluble |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'