missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tris(tetramethylcyclopentadienyl)cerium(III), 98%

Brand:  Alfa Aesar™ 044380.03

Code : NEW

Additional Details : CAS Number : 251984-08-4 Weight : 0.00100kg Transport : UN number : 3181 Chem class : 4.1 Pack group : III

Quantity 1g

Product Code. 15451987

  • £91.90 / 1g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Specifications

Tris (tetramethylcyclopentadienyl)cerium (III)
Powder
C27H39Ce
MFCD01863662
Green
Moisture Sensitive
251984-08-4
98%
503.73
UN3181
1g
Videos
SDS