missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tris(n-propylcyclopentadienyl)yttrium(III)

Brand:  Alfa Aesar™ 039575.03

Code : NEW

Additional Details : CAS Number : 329735-73-1 Weight : 0.00100kg Transport : UN number : 3181 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11384477

  • £83.21 / 1g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Tris(n-propylcyclopentadienyl)yttrium(III)
410.44
UN3181
329735-73-1
MFCD00145488

Specifications

Tris(n-propylcyclopentadienyl)yttrium(III)
Crystalline
MFCD00145488
Pale yellow
1g
329735-73-1
410.44
UN3181
60° to 63°C
Air & Moisture Sensitive
Videos
SDS