missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tris[N,N-bis(trimethylsilyl)amide]neodymium(III), 98%

Brand:  Alfa Aesar™ 044395.03

Code : NEW

Additional Details : CAS Number : 148274-47-9 Weight : 0.00100kg Transport : UN number : 3396 Chem class : 1 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11448590

  • £95.08 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Tris[N,N-bis (trimethylsilyl)amide]neodymium (III)
C18H54N3NdSi6
625.39
UN3396
148274-47-9
[[(CH3)3Si]2N]3Nd
MFCD03427089
Neodymium(III) tris[N,N-bis(trimethylsilyl)amide]

Specifications

Tris[N,N-bis (trimethylsilyl)amide]neodymium (III)
Powder
C18H54N3NdSi6
625.39
UN3396
Pale purple
1g
148274-47-9
98%
[[(CH3)3Si]2N]3Nd
MFCD03427089
Neodymium(III) tris[N,N-bis(trimethylsilyl)amide]
156° to 159°C
Moisture Sensitive
Videos
SDS