missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris[N,N-bis(trimethylsilyl)amide]gadolinium(III), 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Tris[N,N-bis(trimethylsilyl)amide]gadolinium(III) is used in many asymmetric catalysis applications, glove box and Schlenk techniques should be employed to prevent exposure of the rare earth catalyst to air and moisture, which can be detrimental to the reaction outcome.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 161°C to 167°C |
Linear Formula | [[(CH3)3Si]2N]3Gd |
Quantity | 1 g |
UN Number | UN3396 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 638.41 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Tris[N,N-bis (trimethylsilyl)amide]gadolinium (III) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'