missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tris[N,N-bis(trimethylsilyl)amide]europium(III), 98%

Brand:  Alfa Aesar™ 044376.03

Code : NEW

Additional Details : CAS Number : 35789-02-7 Weight : 0.00100kg Transport : UN number : 3396 Chem class : 1 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11468550

  • £101.44 / 1g


Estimated Shipment date
from Supplier 24-04-2018
Add to basket

Specifications

Chemical Identifiers

Tris[N,N-bis (trimethylsilyl)amide]europium (III)
C18H54EuN3Si6
633.13
UN3396
35789-02-7
[[(CH3)3Si]2N]3Eu
MFCD03411248
Europium(III) tris[N,N-bis(trimethylsilyl)amide]

Specifications

Tris[N,N-bis (trimethylsilyl)amide]europium (III)
Powder
C18H54EuN3Si6
633.13
UN3396
Orange
105°C (104mmHg)
Moisture Sensitive
35789-02-7
98%
[[(CH3)3Si]2N]3Eu
MFCD03411248
Europium(III) tris[N,N-bis(trimethylsilyl)amide]
180° to 184°C
1g
Videos
SDS