missing translation for 'onlineSavingsMsg'
Learn More

Tris(n-butylcyclopentadienyl)yttrium(III), 99.9% (REO), Thermo Scientific™

Catalog Number p-7050844
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
312739-77-8
MFCD00145473
This item is not returnable. View return policy
312739-77-8
MFCD00145473

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN1993
Sensitivity Air and moisture sensitive
Formula Weight 452.52
Percent Purity 99.9%
Physical Form Liquid
Assay (REO)
Chemical Name or Material Tris(n-butylcyclopentadienyl)yttrium(III)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'