missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tris(isopropylcyclopentadienyl)neodymium(III), 99.9% (REO)

Brand:  Alfa Aesar™ 039299.03

Code : NEW

Additional Details : CAS Number : 69021-85-8 Weight : 0.00100kg Transport : UN number : 3181 Chem class : 1 Pack group : III

Product Code. 15426305

  • £111.41 / 1g


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Specifications

Tris(isopropylcyclopentadienyl)neodymium(III)
Crystalline
(REO)
465.77
UN3181
Air & Moisture Sensitive
69021-85-8
99.9%
Ampule
MFCD00145483
1g
Videos
SDS