missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tris(isopropylcyclopentadienyl)erbium(III), 99.9% (REO)

Brand:  Alfa Aesar™ 038584.06

Code : NEW

Additional Details : CAS Number : 130521-76-5 Weight : 0.00500kg Transport : UN number : 3181 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11358897

  • £517.49 / 5g


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

Tris(isopropylcyclopentadienyl)erbium(III)
488.79
UN3181
130521-76-5
MFCD00145481

Specifications

Tris(isopropylcyclopentadienyl)erbium(III)
Crystalline
(REO)
488.79
UN3181
5g
130521-76-5
99.9%
Ampule
MFCD00145481
222°C (10mmHg) (subl.)
Air & Moisture Sensitive
Videos
SDS