missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(Hydroxymethyl)Methylamine Hydrochloride, 'Tris-HCl', for Biochemistry, Fisher Chemical™
Click to view available options
Quantity:
100 g
1 kg
Description
Tris(Hydroxymethyl)Methylamine Hydrochloride, 'Tris-HCl', for Biochemistry
Specifications
Melting Point | 150°C |
Color | White |
pH | 3.5 to 5.0 |
Packaging | Plastic powder jar |
Quantity | 1 kg |
Formula Weight | 157.6g/mol |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Tris(hydroxymethyl)aminomethane hydrochloride
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'