missing translation for 'onlineSavingsMsg'
Learn More

Tris(Hydroxymethyl)Methylamine Hydrochloride, 'Tris-HCl', for Biochemistry, Fisher Chemical™

Brand:  Fisher Chemical T/P631/53

Code : 25

Additional Details : CAS Number : 1185-53-1 Weight : 0.50000kg


 View more versions of this product

Product Code. 10142400

  • £332.00 / 500g

In stock, ready to ship: 7

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Tris(Hydroxymethyl)Methylamine Hydrochloride, 'Tris-HCl', for Biochemistry

Specifications

Specifications

Plastic powder jar
150°C
3.5 to 5.0
C4H12ClNO3
tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, tris-hcl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
C(C(CO)(CO)N)O.Cl
157.594
157.6g/mol
White
500g
1185-53-1
12590
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
93573
Solid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tris(hydroxymethyl)aminomethane hydrochloride

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates
View accessory products