missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(ethylenediamine)rhodium(III) chloride trihydrate, Rh 23.2%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
1 g
5 g
Description
Used as precursor for supported RH catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 280°C (decomposition) |
Color | White |
Linear Formula | Rh(H2NCH2CH2NH2)3Cl3·3H2O |
Quantity | 0.5 g |
Formula Weight | 443.61 (389.56 Anhydrous) |
Concentration or Composition (by Analyte or Components) | Rh 23.2% |
Physical Form | Crystalline/Fused Lump, Crystals Fuse Over Time |
Chemical Name or Material | Tris(ethylenediamine)rhodium(III) chloride trihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'