Learn More
TE Buffer, Tris-EDTA, 10X Solution, pH 7.6, Molecular Biology, Fisher BioReagents
Brand: Fisher Bioreagents BP2475-500
Code : 32
Additional Details : CAS Number : 38641-82-6 Weight : 0.55000kg
Quantity | 500mL |
---|---|
Packaging | Poly Bottle |
Description
- Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TE buffer solution - simply dilute as needed
- 10X solution contains 100mM tris and 10mM EDTA
- To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TE solution with 9 volumes of ultrapure water, such as BP2819
- Filtered through a 5 micron filter and autoclaved
- Tested for the absence of DNase, RNase, and protease
- TE buffer is routinely used for resuspending nucleic acid samples
- Intended for laboratory research use only
Chemical Identifiers
Tris-EDTA (TE), 10X Solution | |
7732-18-5 | |
TE |
pH 7.6 | |
Molecular Biology Grade |
Specifications
Tris-EDTA (TE), 10X Solution | |
7732-18-5 | |
Liquid | |
TE | |
Protease free | |
7.4 to 8.1 | |
500mL |
pH 7.6 | |
Molecular Biology Grade | |
DNase free | |
Poly Bottle | |
Filtered through a 5-micron filter and autoclaved. | |
Colorless |