missing translation for 'onlineSavingsMsg'
Learn More

Tris(dioxa-3,6-heptyl)amine, 95%, Thermo Scientific Chemicals

Catalog Number 10512521
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
70384-51-9
C15H33NO6
323.42
MFCD00010748
XGLVDUUYFKXKPL-UHFFFAOYSA-N
tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine
112414
2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine
COCCOCCN(CCOCCOC)CCOCCOC
This item is not returnable. View return policy
70384-51-9
C15H33NO6
323.42
MFCD00010748
XGLVDUUYFKXKPL-UHFFFAOYSA-N
tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine
112414
2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine
COCCOCCN(CCOCCOC)CCOCCOC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Density 1.0100g/mL
Boiling Point 330.0°C
Flash Point 162°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (CH3OCH2CH2OCH2CH2)3N
Refractive Index 1.4466 to 1.4486
Quantity 25 mL
Fieser 15,356; 16,373
Specific Gravity 1.01
Solubility Information Solubility in water: miscible. Other solubilities: soluble in acetone, ether, toluene and cyclohexane
Formula Weight 323.42
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tris(dioxa-3, 6-heptyl)amine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'