missing translation for 'onlineSavingsMsg'
Learn More

Tris(dioxa-3,6-heptyl)amine, 95%, ACROS Organics™

Brand:  Acros Organics 124855000

Code : 29

Additional Details : CAS Number : 70384-51-9 Weight : 0.55500kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 500mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10465675

  • £389.20 / 500mL


Estimated Shipment
13-02-2018
Add to basket

Specifications

Chemical Identifiers

Tris(dioxa-3, 6-heptyl)amine
95%
(CH3OCH2CH2OCH2CH2)3N
MFCD00010748
95%
C15H33NO6
323.42
TDA-1

Specifications

Tris(dioxa-3, 6-heptyl)amine
Authentic
95%
C15H33NO6
Glass bottle
MFCD00010748
Solubility in water: miscible. Other solubilities: soluble in acetone, ether, toluene and cyclohexane
1.01
Clear pale yellow to brown
162°C
70384-51-9
Liquid
95%
(CH3OCH2CH2OCH2CH2)3N
323.42
1.4466 to 1.4486
TDA-1
1.0100g/mL
330.0°C
500mL
Videos
SDS