missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform, Pd 20.6%

Brand:  Alfa Aesar™ 044969.03

Code : NEW

Additional Details : CAS Number : 52522-40-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416655

  • £66.50 / 1g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Tris (dibenzylideneacetone)dipalladium (0)
Complex with chloroform
Pd2(C17H14O)3·CHCl3
MFCD00075479
52522-40-4
C51H42O3Pd2·CHCl3
1035.11

Specifications

Tris (dibenzylideneacetone)dipalladium (0)
52522-40-4
C51H42O3Pd2·CHCl3
1035.11
131° to 135°C
1g
Complex with chloroform
Powder
Pd2(C17H14O)3·CHCl3
MFCD00075479
Pd 20.6%
Air & Moisture Sensitive
Videos
SDS