missing translation for 'onlineSavingsMsg'
Learn More

Tris(cyclopentadienyl)yttrium(III), 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7049476
55.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1294-07-1
C15H15Y
284.191
MFCD00058706
IUJOPJIXAVSZIQ-UHFFFAOYSA-N
yttrium 3+ ion tris cyclopenta-1,3-dien-1-ide
22481132
cyclopenta-1,3-diene;yttrium(3+)
C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Y+3]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1294-07-1
C15H15Y
284.191
MFCD00058706
IUJOPJIXAVSZIQ-UHFFFAOYSA-N
yttrium 3+ ion tris cyclopenta-1,3-dien-1-ide
22481132
cyclopenta-1,3-diene;yttrium(3+)
C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Y+3]

Tris(cyclopentadienyl)yttrium(III) is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 295°C
Boiling Point 200°C (sublimation)(vacuum)
Packaging Ampule
Quantity 0.5 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 284.2
Percent Purity 99.9%
Physical Form Powder
Assay (REO)
Chemical Name or Material Tris(cyclopentadienyl)yttrium(III)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'