missing translation for 'onlineSavingsMsg'
Learn More

Tris(cyclopentadienyl)gadolinium(III), 99.9% (REO), Thermo Scientific™

Catalog Number p-7050547
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1272-21-5
C15H15Gd
352.535
MFCD00145476
WPVCUPCREAIUBK-UHFFFAOYSA-N
tris cyclopentadienyl gadolinium-gd reo
73411059
cyclopenta-1,3-diene;gadolinium(3+)
C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Gd+3]
This item is not returnable. View return policy
1272-21-5
C15H15Gd
352.535
MFCD00145476
WPVCUPCREAIUBK-UHFFFAOYSA-N
tris cyclopentadienyl gadolinium-gd reo
73411059
cyclopenta-1,3-diene;gadolinium(3+)
C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Gd+3]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Packaging Ampule
Quantity 1 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Formula Weight 352.54
Percent Purity 99.9%
Physical Form Powder
Assay (REO)
Chemical Name or Material Tris(cyclopentadienyl)gadolinium(III)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'