missing translation for 'onlineSavingsMsg'
Learn More

TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™

Catalog Number p-29103
51.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


45.43 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


62.81 GBP valid until 2024-12-31
Use promo code "23286" to get your promotional price.


Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Nalgene Poly Bottle
Poly CUBE
10043-35-3, 60-00-4, 77-86-1, 7732-18-5
TBE
10043-35-3, 60-00-4, 77-86-1, 7732-18-5
TBE

TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TBE buffer solution - simply dilute as needed
  • 10X solution contains 0.89M tris, 0.89M borate, and 0.02M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBE Solution with 9 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Functionally tested for use in electrophoresis applications
  • TBE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY
Color Colorless
pH 8.2 to 8.4
Physical Form Liquid
Quantity 1 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Packaging Nalgene Poly Bottle
Protease Protease free
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Tris-Borate-EDTA, 10X Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'