missing translation for 'onlineSavingsMsg'
Learn More

TRIS (Base), Ultrapure Bioreagent, J.T. Baker™

Catalog Number p-5391313
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Molecular Biology Applications
Packaging Poly Bottle
Quantity 1 kg
Formula Weight 121.14
Grade Ultrapure Bioreagent
Chemical Name or Material TRIS (Base)
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'