missing translation for 'onlineSavingsMsg'
Learn More

TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents

Brand:  Fisher Bioreagents BP1332-1

Code : 32

Additional Details : CAS Number : 135852-26-5 Weight : 1.05000kg

Quantity 1L
Packaging Nalgene Poly Bottle

 View more versions of this product

Product Code. 10490264

  • £73.54 / 1 L


Estimated Shipment
04-05-2018
Add to basket

Description

Description

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 50X TAE Solution - simply dilute as needed!
  • 50X solution contains 2M tris-acetate and 0.050M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 50X TAE
  • Solution with 49 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only
Specifications

Chemical Identifiers

Tris-Acetate-EDTA
6850-28-8
DNase-, RNase- and Protease-Free
TAE
50X Solution
Electrophoresis Grade
Liquid

Specifications

Tris-Acetate-EDTA
6850-28-8
DNase-, RNase- and Protease-Free
DNase free
TAE
Nalgene Poly Bottle
Filtered through a 0.2-micron filter.
50X Solution
Electrophoresis Grade
Liquid
Pass Test
Tris (24%), Acetic Acid (5.0%), and EDTA (<2%).
Protease free
1L
Videos
SDS Documentation