missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris, 99.8 to 100.1% (Dry Basis), Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
Packaging:
Plastic bottle
Specifications
Melting Point | 171°C to 172°C |
Boiling Point | 219°C to 220°C (10 mmHg) |
Physical Form | Crystalline Powder |
Quantity | 500 g |
Linear Formula | NH2C(CH2OH)3 |
Packaging | Plastic bottle |
Beilstein | 741883 |
Formula Weight | 121.14 |
Percent Purity | 99.8 to 100.1% |
Grade | Molecular Biology |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'