Learn More
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium(III), 99% (metals basis), Thermo Scientific Chemicals
Description
It is a reactant involved in the synthesis of rare earth metal complexes for separation of lanthanide metals, conformational mobility of substituted 2-methoxychalcones under the action of lanthanide shift reagents, NMR titration on cation-π complexes of bowl-shaped polycyclic aromatic hydrocarbons, specific recognition of chiral amino alcohols. It finds its application as catalysis and synthesis, optics & glasses, healthcare & biochemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 108°C to 111°C |
Quantity | 1 g |
Sensitivity | Hygroscopic |
Solubility Information | Slowly degrades in water and soluble in hydrocarbons,alcohols,ketones,esters. |
Formula Weight | 1058.59 |
Percent Purity | 99% |
Assay | (metals basis) |
Chemical Name or Material | Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium(III) |
RUO – Research Use Only