missing translation for 'onlineSavingsMsg'
Learn More

Tris(4,4,4-trifluoro-1-(2-thienyl)-1,3-butanediono)europium(III) hydrate, Thermo Scientific Chemicals

Catalog Number p-7058845
Click to view available options
Quantity:
0.1 g
0.5 g
This item is not returnable. View return policy
41884-77-9
C24H12EuF9O6S3
815.48
MFCD00041948
JIYALKNSIOHINA-UHFFFAOYSA-N
Europium(III) thenoyltrifluoroacetonate hydrate
europium(3+) tris(4,4,4-trifluoro-1,3-dioxo-1-(thiophen-2-yl)butan-2-ide)
[Eu+3].FC(F)(F)C(=O)[CH-]C(=O)C1=CC=CS1.FC(F)(F)C(=O)[CH-]C(=O)C1=CC=CS1.FC(F)(F)C(=O)[CH-]C(=O)C1=CC=CS1
This item is not returnable. View return policy
41884-77-9
C24H12EuF9O6S3
815.48
MFCD00041948
JIYALKNSIOHINA-UHFFFAOYSA-N
Europium(III) thenoyltrifluoroacetonate hydrate
europium(3+) tris(4,4,4-trifluoro-1,3-dioxo-1-(thiophen-2-yl)butan-2-ide)
[Eu+3].FC(F)(F)C(=O)[CH-]C(=O)C1=CC=CS1.FC(F)(F)C(=O)[CH-]C(=O)C1=CC=CS1.FC(F)(F)C(=O)[CH-]C(=O)C1=CC=CS1

As a laser dye

Tris(4,4,4-trifluoro-1-(2-thienyl)-1,3-butanediono)europium(III) hydrate is used as a laser dye. It acts as a chelator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 143°C to 144°C
Color Yellow
Quantity 0.1 g
Solubility Information Soluble in methanol, toluene
Formula Weight 815.51 (Anhydrous)
Physical Form Powder
Chemical Name or Material Tris (4,4,4-trifluoro-1- (2-thienyl)-1,3-butanediono)europium (III) hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'