missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris (2-cyanoethyl)phosphine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
10 g
50 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 97.0°C to 99.0°C |
Boiling Point | 235.0°C (0.9 mmHg) |
Assay Percent Range | 94% min. (Iodimetry) |
Packaging | Glass bottle |
Quantity | 10 g |
Solubility Information | Solubility in water: slightly soluble |
Formula Weight | 193.19 |
Percent Purity | 95% |
Chemical Name or Material | Tris (2-cyanoethyl)phosphine |
Safety and Handling
Product Identifier
- Tris (2-cyanoethyl)phosphine
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H350-May cause cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- EUH208-Contains . May produce an allergic reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- P313-Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'