missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-aminoethyl)amine, 96%, Thermo Scientific Chemicals

Catalog Number p-4510543
137.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
4097-89-6
C6H18N4
146.24
MBYLVOKEDDQJDY-UHFFFAOYSA-N
tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine
77731
CHEBI:30631
N',N'-bis(2-aminoethyl)ethane-1,2-diamine
C(CN(CCN)CCN)N
4097-89-6
C6H18N4
146.24
MBYLVOKEDDQJDY-UHFFFAOYSA-N
tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine
77731
CHEBI:30631
N',N'-bis(2-aminoethyl)ethane-1,2-diamine
C(CN(CCN)CCN)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -16.0°C
Color Yellow
Density 0.9770g/mL
Boiling Point 265.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula (H2NCH2CH2)3N
Refractive Index 1.4960 to 1.4980
Quantity 100 mL
Beilstein 04,II,695
Specific Gravity 0.977
Solubility Information Solubility in water: soluble.
Formula Weight 146.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tris(2-aminoethyl)amine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'