missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III), recrystallized, 99.9+% (metals basis)

Brand:  Alfa Aesar™ 044003.14

Code : NEW

Additional Details : CAS Number : 15632-39-0 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11325078

  • £613.30 / 25g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium (III)
Recrystallized
638.7
Yttrium(III)-DPM; Y(TMHD)3
15632-39-0
Y[(CH3)3CCOCH=COC(CH3)3]3
MFCD00015713

Specifications

Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium (III)
15632-39-0
≥99.9%
Y[(CH3)3CCOCH=COC(CH3)3]3
MFCD00015713
170° to 173°C
25g
Recrystallized
Crystalline
(metals basis)
638.7
Yttrium(III)-DPM; Y(TMHD)3
95°C (0.05mmHg)
Videos
SDS