missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)scandium(III), 95+%, Thermo Scientific™

Catalog Number p-7058581
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy
15492-49-6
C33H57O6Sc
594.769
MFCD00169875
SIPNKDOCYGGAQY-LWTKGLMZSA-K
scandium iii 2,2,6,6-tetramethyl-3,5-heptanedionate, tris 2,2,6,6-tetramethyl-3,5-heptanedionato scandium, 4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy scandio oxy-2,2,6,6-tetramethylhept-4-en-3-one, scandium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate hydrate trace metals basis, tris 2,2,6,6-tetramethyl-3,5-heptanedionato scandium iii-sc reo sc tmhd 3
134068149
scandium(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Sc+3]
This item is not returnable. View return policy
15492-49-6
C33H57O6Sc
594.769
MFCD00169875
SIPNKDOCYGGAQY-LWTKGLMZSA-K
scandium iii 2,2,6,6-tetramethyl-3,5-heptanedionate, tris 2,2,6,6-tetramethyl-3,5-heptanedionato scandium, 4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy scandio oxy-2,2,6,6-tetramethylhept-4-en-3-one, scandium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate hydrate trace metals basis, tris 2,2,6,6-tetramethyl-3,5-heptanedionato scandium iii-sc reo sc tmhd 3
134068149
scandium(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Sc+3]

Intermediate for scandium oxide ceramics

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 150°C to 152°C
Color White
Boiling Point 275°C (decomposition)
Flash Point >110°C (230°F)
Quantity 0.5 g
Sensitivity Hygroscopic
Solubility Information Soluble in ethyl acetate
Formula Weight 594.77
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)scandium (III)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'