missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium(III), 99%, Thermo Scientific™

Catalog Number p-7082027
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15492-48-5
C33H57O6Pr
690.72
MFCD00000027,MFCD00000027
HDONDLBRVFKILD-UHFFFAOYSA-K
pr dpm 3, praseodymium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate
53384607
praseodymium(3+);(E)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
[Pr+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C
This item is not returnable. View return policy
15492-48-5
C33H57O6Pr
690.72
MFCD00000027,MFCD00000027
HDONDLBRVFKILD-UHFFFAOYSA-K
pr dpm 3, praseodymium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate
53384607
praseodymium(3+);(E)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
[Pr+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 219°C to 221°C
Quantity 1 g
Formula Weight 690.72
Percent Purity 99%
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium (III)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'