Learn More
Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)neodymium(III)
Brand:  Alfa Aesar™ 039298.06
Code : NEW
Additional Details : CAS Number : 15492-47-4 Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 11383698
  • £129.00 / 5g
Estimated Shipment date
from Supplier 11-06-2018
Add to basket
Specifications
Chemical Identifiers
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)neodymium(III)
694.06
Neodymium(III)-DPM; Nd(TMHD)3
15492-47-4
MFCD00015708
Specifications
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)neodymium(III)
Crystalline
MFCD00015708
209° to 212°C
5g
15492-47-4
694.06
Neodymium(III)-DPM; Nd(TMHD)3
270°C (dec.)
Hygroscopic