missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gallium(III)

Brand:  Alfa Aesar™ 014491.03

Code : NEW

Additional Details : CAS Number : 34228-15-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11312448

  • £73.25 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gallium(III)
619.54
Gallium(III)-DPM; Ga(TMHD)3
34228-15-4
MFCD00058705

Specifications

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gallium(III)
Crystalline
MFCD00058705
White
1g
34228-15-4
619.54
Gallium(III)-DPM; Ga(TMHD)3
219° to 220°C
Hygroscopic
Videos
SDS