missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III), 98%

Superconductor MOCVD Precursor

Brand:  Alfa Aesar™ 036464.04

Code : NEW

Additional Details : CAS Number : 15522-69-7 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11307928

  • £55.90 / 2g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III)
712.32
Dysprosium(III)-DPM; Dy(TMHD)3
15522-69-7
MFCD00000017

Specifications

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III)
Powder
712.32
Dysprosium(III)-DPM; Dy(TMHD)3
2g
15522-69-7
98%
MFCD00000017
182° to 183°C
Hygroscopic
Videos
SDS