missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99.9% (metals basis), Thermo Scientific™

Catalog Number p-7051472
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14877-41-9
C33H57CoO6
608.75
MFCD00233617,MFCD00233617
UJRJNARBHKKZST-UHFFFAOYSA-K
cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
131675252
cobalt(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
[Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C
This item is not returnable. View return policy
14877-41-9
C33H57CoO6
608.75
MFCD00233617,MFCD00233617
UJRJNARBHKKZST-UHFFFAOYSA-K
cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
131675252
cobalt(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
[Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 254°C to 256°C
Color Green
Boiling Point 120°C (0.5 mmHg)(sublimation)
Odor Odorless
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 608.72
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'